Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant Rat MCP-1(MCAF). Anti-Rat MCP-1(MCAF) specific antib ody was purified by affinity chromatography employing immobilized Rat MCP-1(MCAF) matrix. 1 * 1 mg
Catalog # PEPR500-P76-1
Supplier: PeproTech, Inc.
Frequently Bought Together