Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mMDC. Anti-Murine MDC specific antibody was purified by affinity chromatography employing immobilized mMDC matrix. 1 * 1 mg
PEPR500-P176-1
Frequently Bought Together