ContactUsLinkComponent
Produced from sera of goats pre-immunized with highly pure (>98%) recombinant hEotaxin-3. Anti-Human Eotaxin-3 specific antibody was purified by affinity chromatography employing immobilized hEotaxin-3 matrix. 1 * 1 mg
Katalognr. PEPR500-P156G-1
Supplier:  PeproTech, Inc.
Produced from sera of goats pre-immunized with highly pure (>98%) recombinant hEotaxin-3. Anti-Human Eotaxin-3 specific antibody was purified by affinity chromatography employing immobilized hEotaxin-3 matrix. 1 * 1 mg
Katalognr. PEPR500-P156G-1
Supplier:  PeproTech, Inc.
Frequently Bought Together