ContactUsLinkComponent
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hFGF-17. Anti-Human FGF-17 specific antibody was pur ified by affinity chromatography employing immobilized hFGF-17 matrix. 1 * 1 mg
Catalog # PEPR500-P152-1
Supplier:  PeproTech, Inc.
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hFGF-17. Anti-Human FGF-17 specific antibody was pur ified by affinity chromatography employing immobilized hFGF-17 matrix. 1 * 1 mg
Catalog # PEPR500-P152-1
Supplier:  PeproTech, Inc.
Frequently Bought Together