ContactUsLinkComponent
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hsTNF-receptor. Anti-Human sTNF-receptor specific an tibody was purified by affinity chromatography employing immobilized hsTNF-receptor matrix. 1 * 1 mg
Référence PEPR500-P143-1
Fournisseur:  PeproTech, Inc.
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hsTNF-receptor. Anti-Human sTNF-receptor specific an tibody was purified by affinity chromatography employing immobilized hsTNF-receptor matrix. 1 * 1 mg
Référence PEPR500-P143-1
Fournisseur:  PeproTech, Inc.
Frequently Bought Together