ContactUsLinkComponent
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mJE(MCP-1). Anti-Murine JE(MCP-1) specific antibody was purified by affinity chromatography employing immobilized mJE (MCP-1) matrix. 1 * 1 mg
Référence PEPR500-P113-1
Fournisseur:  PeproTech, Inc.
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mJE(MCP-1). Anti-Murine JE(MCP-1) specific antibody was purified by affinity chromatography employing immobilized mJE (MCP-1) matrix. 1 * 1 mg
Référence PEPR500-P113-1
Fournisseur:  PeproTech, Inc.
Frequently Bought Together