Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mC10. Anti-Murine C10 specific antibody was purified by affinity chromatography employing immobilized mC10 matrix. 1 * 1 mg
Catalog # PEPR500-P112-1
Fournisseur: PeproTech, Inc.
Frequently Bought Together