ContactUsLinkComponent
Synonyms: Macrophage-Derived Chemokine CCL22 STCP-1 ABCD-1 Description: MDC is a CC chemokine that is produced in B cells, macrop hages, monocyte-derived dendritic cells, activated NK cells and CD4 T cells. It signals through the CCR4 receptor. MD 1 * 1 mg
  PEPR300-36A-1
Synonyms: Macrophage-Derived Chemokine CCL22 STCP-1 ABCD-1 Description: MDC is a CC chemokine that is produced in B cells, macrop hages, monocyte-derived dendritic cells, activated NK cells and CD4 T cells. It signals through the CCR4 receptor. MD 1 * 1 mg
  PEPR300-36A-1
Frequently Bought Together