Biotin NTA
Biotin NTA
Biotin NTA or BNTA (Biotin nitrilotriacetic acid, tripotassium salt), 5 mg
Catalog # 90074.
Supplier:   Biotium
Biotin NTA
Catalog # 90074.
Supplier:  Biotium
CAS Number:  
Biotin NTA
Biotin NTA
Biotin NTA or BNTA (Biotin nitrilotriacetic acid, tripotassium salt), 5 mg
CAS Number:  
Price
SFr. 237.00
Log in for availability
1 * 5 mg
Product Details & Documents
Biotinylated probes for detection of His-tagged proteins immobilised on nitrocellulose membranes.
  • White solid soluble in water
  • Removable from the histidine-tagged protein at pH 4,8
Biotin NTA, also known as BNTA (Biotin nitrilotriacetic acid, tripotassium salt) reversibly binds to histidine-tagged proteins, and can be detected with Biotium's CF™ Dye streptavidin or monoclonal anti-biotin fluorescent conjugates. Available with 6-atom aminohexanoyl (X) linker between the biotin and NTA to facilitate interaction with streptavidin.
Specifications
Specifications
  • Cdt:
    5 mg
  • Désignation:
    Biotin NTA or BNTA (Biotin nitrilotriacetic acid, tripotassium salt)
Specifications
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..