Software for data loggers, elproLOG ANALYZE
elproLOG ANALYZE software, update
N° de catalogue ELPR800396
Fournisseur: ELPRO-BUCHS
Aller à
Product Details & Documents
elproLOG ANALYZE is a software for all elpro data loggers. It provides all functions such as data analysis, reporting and configuration of data loggers.
- User friendly
- Displays statistics on measurement data
- Optional audit trail
- Multiple zoom function
Spécifications
- Désignation:elproLOG ANALYZE software, update
- Seq:02