Bains cryothermostats circulation, srie Optima R, Grant, Ouverture l x p: 110x145 mm, Niveau de liquide min/max: 80/140 mm, GR1 50-R2, Gamme de temp.: -20...100 C, Prcision de temp.: 0.1 C 1 * 1 Pce
Catalog # 461-0219
Supplier: GRANT INSTRUMENTS
Frequently Bought Together