Double-spatules, larges
500 mm, 30 mm
N° de catalogue RSGA092.500
Fournisseur: RSG
Aller à
Product Details & Documents
Double spatulas with wide, flat endings. Made of stainless steel.
Spécifications
- Longueur:500 mm
- Largeur de la lame:30 mm
Double spatulas with wide, flat endings. Made of stainless steel.