Microspatules doubles, rigides
Microspatules doubles, rigides
130 mm, 3 mm
Catalog # RSGA833.130
Supplier:   RSG
Microspatules doubles, rigides
Catalog # RSGA833.130
Supplier:  RSG
Microspatules doubles, rigides
Microspatules doubles, rigides
130 mm, 3 mm
Price
SFr. 28.50
Log in for availability
1 * 5 ST
Product Details & Documents
Rigid micro double spatulas made of stainless steel.
Specifications
  • Longueur:
    130 mm
  • Largeur de la lame:
    3 mm