Microtitre tubes, 1,2 ml
Microtitre tubes, 1,2 ml, incolore, Bulk
N° de catalogue BZYM710410
Marketsource
Fournisseur: BIOZYM
Aller à
Product Details & Documents
These standard 1,2 ml microtiter tubes are made of polypropylene and have a labeling field.
- Autoclavable
- Freezable
- Can be closed with PlugStrip cap strips
The standard racks can be refilled easily and inexpensively with the reload system. Rack version works on Beckman, Tecan and Tomtec systems.
For individual vessels, the PlugStrip closures can be individually tailored. All microtiter tubes have an additional filling space that allows sample dilutions and/or sample mixing prior to transfer to microplates.
Possible applications include: HTLV-III tests, RIA, transport of specimen and reagents.
Ordering information: Available in sachets as well as pre-fitted in 8×12 format. Also available is a robot-compatible rack version.
Spécifications
- Emballage:Bulk
- Capacité:1,2 ml
- Couleur:incolore
- Désignation:Microtitre tubes
- Seq:01



