Microtubes à centrifuger
1,6 ml, naturel, with attached cap, Graduated, conical bottom, non sterile, pyrogene free, 20000×g, 1000 pieces/bag
N° de catalogue BZYM710340
Marketsource
Fournisseur: BIOZYM
Aller à
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Certificats
Rechercher par numéro de lotSpécifications
- Emballage:1000 pieces/bag
- Type:with attached cap
- Version:Graduated, conical bottom, non sterile, pyrogene free
- Capacité:1,6 ml
- Couleur:naturel
- Seq:39
- RCF max.:20000×g