Microtubes à centrifuger
2,0 ml, naturel, With attached cap, Graduated, conical bottom, non sterile, RNAse free, 20000×g, 500 pieces/bag
N° de catalogue BZYM710320
Marketsource
Fournisseur: BIOZYM
Aller à
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Documents
Certificats
Rechercher par numéro de lotSpécifications
- Emballage:500 pieces/bag
- Type:With attached cap
- Version:Graduated, conical bottom, non sterile, RNAse free
- Capacité:2,0 ml
- Couleur:naturel
- Seq:49
- RCF max.:20000×g