Microtubes à centrifuger
2,0 ml, assortiment, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 500 pieces/bag
Catalog # BZYM710198
Marketsource
Supplier:   BIOZYM
Microtubes à centrifuger
Catalog # BZYM710198
Supplier:  BIOZYM
Marketsource
Microtubes à centrifuger
2,0 ml, assortiment, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 500 pieces/bag
Price
SFr. 21.07
Log in for availability
1 * 500 ST
Product Details & Documents
PE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • DNase-, RNase free
With or without cap.
Specifications
  • Emballage:
    500 pieces/bag
  • Type:
    With attached cap
  • Version:
    Graduated, conical bottom, non sterile
  • Capacité:
    2,0 ml
  • Couleur:
    assortiment
  • Seq:
    47
  • RCF max.:
    20000×g