Microtubes à centrifuger
2,0 ml, assortiment, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 500 pieces/bag
Catalog # BZYM710198
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Certificates
Search by Lot NumberSpecifications
- Emballage:500 pieces/bag
- Type:With attached cap
- Version:Graduated, conical bottom, non sterile
- Capacité:2,0 ml
- Couleur:assortiment
- Seq:47
- RCF max.:20000×g