Microtubes à centrifuger
1,6 ml, bleu, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 500 pieces/bag
BZYM710162
Marketsource
: BIOZYM
- Emballage:500 pieces/bag
- Type:With attached cap
- Version:Graduated, conical bottom, non sterile
- Capacité:1,6 ml
- Couleur:bleu
- Seq:27
- RCF max.:20000×g