Microtubes à centrifuger
Microtubes à centrifuger
1,6 ml, rouge, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 500 pieces/bag
Katalognr. BZYM710161
Marketsource
Anbieter:   BIOZYM
Microtubes à centrifuger
Katalognr. BZYM710161
Anbieter:  BIOZYM
Marketsource
Microtubes à centrifuger
1,6 ml, rouge, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 500 pieces/bag
Preis
SFr. 18.14
Einloggen für Verfügbarkeit
1 * 500 ST
Product Details & Documents
PE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • DNase-, RNase free
With or without cap.
Technische Daten
  • Emballage:
    500 pieces/bag
  • Type:
    With attached cap
  • Version:
    Graduated, conical bottom, non sterile
  • Capacité:
    1,6 ml
  • Couleur:
    rouge
  • Seq:
    31
  • RCF max.:
    20000×g