Microtubes à centrifuger
1,6 ml, naturel, With attached cap, Non graduated, conical bottom, non sterile, 20000×g, 500 pieces/bag
Catalog # BZYM710157
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Certificates
Search by Lot NumberSpecifications
- Emballage:500 pieces/bag
- Type:With attached cap
- Version:Non graduated, conical bottom, non sterile
- Capacité:1,6 ml
- Couleur:naturel
- Seq:35
- RCF max.:20000×g