Microtubes à centrifuger
0,6 ml, naturel, with attached cap, Graduated, conical bottom, sterile, 20000×g, 500 pieces/bag (individually wrapped )
Catalog # BZYM710135
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Documents
Certificates
Search by Lot NumberSpecifications
- Emballage:500 pieces/bag (individually wrapped )
- Type:with attached cap
- Version:Graduated, conical bottom, sterile
- Capacité:0,6 ml
- Couleur:naturel
- Seq:04
- RCF max.:20000×g