Microtubes à centrifuger
Microtubes à centrifuger
0,6 ml, naturel, with attached cap, Graduated, conical bottom, sterile, 20000×g, 500 pieces/bag (individually wrapped )
Catalog # BZYM710135
Marketsource
Supplier:   BIOZYM
Microtubes à centrifuger
Catalog # BZYM710135
Supplier:  BIOZYM
Marketsource
Microtubes à centrifuger
0,6 ml, naturel, with attached cap, Graduated, conical bottom, sterile, 20000×g, 500 pieces/bag (individually wrapped )
Price
SFr. 49.65
Log in for availability
1 * 500 ST
Product Details & Documents
PE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • DNase-, RNase free
With or without cap.
Documents
Specifications
  • Emballage:
    500 pieces/bag (individually wrapped )
  • Type:
    with attached cap
  • Version:
    Graduated, conical bottom, sterile
  • Capacité:
    0,6 ml
  • Couleur:
    naturel
  • Seq:
    04
  • RCF max.:
    20000×g