Microtubes à centrifuger
0,6 ml, assortiment de couleurs, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 1000 pieces/bag
BZYM710128
Marketsource
: BIOZYM
- Emballage:1000 pieces/bag
- Type:With attached cap
- Version:Graduated, conical bottom, non sterile
- Capacité:0,6 ml
- Couleur:assortiment de couleurs
- Seq:13
- RCF max.:20000×g