Microtubes à centrifuger
0,6 ml, violet, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 1000 pieces/bag
BZYM710126
Marketsource
: BIOZYM
- Emballage:1000 pieces/bag
- Type:With attached cap
- Version:Graduated, conical bottom, non sterile
- Capacité:0,6 ml
- Couleur:violet
- Seq:11
- RCF max.:20000×g