Microtubes à centrifuger
0,6 ml, vert, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 1000 pieces/bag
  BZYM710123
Marketsource
 :   BIOZYM
Microtubes à centrifuger
  BZYM710123
 :  BIOZYM
Marketsource
Microtubes à centrifuger
0,6 ml, vert, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 1000 pieces/bag
SFr. 32.89
1 * 1.000 ST
PE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • DNase-, RNase free
With or without cap.
  • Emballage:
    1000 pieces/bag
  • Type:
    With attached cap
  • Version:
    Graduated, conical bottom, non sterile
  • Capacité:
    0,6 ml
  • Couleur:
    vert
  • Seq:
    07
  • RCF max.:
    20000×g