Microtubes à centrifuger
0,6 ml, bleu, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 1000 pieces/bag
Catalog # BZYM710122
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Documents
Certificates
Search by Lot NumberSpecifications
- Emballage:1000 pieces/bag
- Type:With attached cap
- Version:Graduated, conical bottom, non sterile
- Capacité:0,6 ml
- Couleur:bleu
- Seq:06
- RCF max.:20000×g