Microtubes à centrifuger
Microtubes à centrifuger
0,6 ml, bleu, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 1000 pieces/bag
Catalog # BZYM710122
Marketsource
Supplier:   BIOZYM
Microtubes à centrifuger
Catalog # BZYM710122
Supplier:  BIOZYM
Marketsource
Microtubes à centrifuger
0,6 ml, bleu, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 1000 pieces/bag
Price
SFr. 32.89
Log in for availability
1 * 1.000 ST
Product Details & Documents
PE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • DNase-, RNase free
With or without cap.
Documents
Specifications
  • Emballage:
    1000 pieces/bag
  • Type:
    With attached cap
  • Version:
    Graduated, conical bottom, non sterile
  • Capacité:
    0,6 ml
  • Couleur:
    bleu
  • Seq:
    06
  • RCF max.:
    20000×g