Microtubes à centrifuger
0,6 ml, jaune, With attached cap, Non graduated, conical bottom, non sterile, 20000×g, 1000 pieces/bag
N° de catalogue BZYM710104
Marketsource
Fournisseur: BIOZYM
Aller à
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Documents
Certificats
Rechercher par numéro de lotSpécifications
- Emballage:1000 pieces/bag
- Type:With attached cap
- Version:Non graduated, conical bottom, non sterile
- Capacité:0,6 ml
- Couleur:jaune
- Seq:20
- RCF max.:20000×g