Microtubes à centrifuger
0,6 ml, vert, With attached cap, Non graduated, conical bottom, non sterile, 20000×g, 1000 pieces/bag
BZYM710103
Marketsource
: BIOZYM
- Emballage:1000 pieces/bag
- Type:With attached cap
- Version:Non graduated, conical bottom, non sterile
- Capacité:0,6 ml
- Couleur:vert
- Seq:15
- RCF max.:20000×g