Sample tubes
1,5 ml, Free standing, incolore, with side label, PP, screw cap with attachment loop, Stérile
N° de catalogue BZYM710066
Marketsource
Fournisseur: BIOZYM
Aller à
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Spécifications
- Stérilité:Stérile
- Type:screw cap with attachment loop
- Capacité:1,5 ml
- Couleur:incolore
- Désignation:with side label
- Matière:PP
- Seq:20
- Fond:Free standing