Sample tubes
1,5 ml, Conical bottom, incolore, with side label, PP, screw cap without attachment loop, Stérile
Catalog # BZYM710052
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Specifications
- Stérilité:Stérile
- Type:screw cap without attachment loop
- Capacité:1,5 ml
- Couleur:incolore
- Désignation:with side label
- Matière:PP
- Seq:17
- Fond:Conical bottom
- Cat. No.:BZYM710052