Sample tubes
1,5 ml, Free standing, incolore, with side label, PP
BZYM710020
Marketsource
: BIOZYM
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
- Capacité:1,5 ml
- Couleur:incolore
- Désignation:with side label
- Matière:PP
- Seq:07
- Fond:Free standing