Sample tubes
0,5 ml, Free standing, incolore, with side label, PP
N° de catalogue BZYM710015
Marketsource
Fournisseur: BIOZYM
Aller à
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Spécifications
- Capacité:0,5 ml
- Couleur:incolore
- Désignation:with side label
- Matière:PP
- Seq:03
- Fond:Free standing