Sample tubes
2,0 ml, Conical bottom, incolore, with side label, PP
BZYM710011
Marketsource
: BIOZYM
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
- Capacité:2,0 ml
- Couleur:incolore
- Désignation:with side label
- Matière:PP
- Seq:09
- Fond:Conical bottom