Sample tubes
1,5 ml, Conical bottom, ambré, with side label, PP
Katalognr. BZYM710006
Marketsource
Anbieter: BIOZYM
Direkt zu
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Technische Daten
- Capacité:1,5 ml
- Couleur:ambré
- Désignation:with side label
- Matière:PP
- Seq:06
- Fond:Conical bottom