Potassium hydroxyde, qualité Réactif
Flacon Plastique, 500 g
: Potash caustic, Caustic potash
- :1310-58-3
- :KOH
- :MFCD00003553
- :56.11 g/mol
- :12,07806
- :1320 °C (1013 hPa)
- :360 °C
- :2,04 g/cm³ (20 °C)
- :Température ambiante
- :215-181-3
- :1813
- :8,II
- :01-2119487136-33
- Arsenic:≤0.0003%
- Heavy Metals:≤0.003%
- Moisture:10 – 17%
- Potassium Carbonate:≤3.5%
- Purity:>85.0%
- Type de conditionnement:Flacon Plastique
- Cdt:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



