Bouchons compte-gouttes, WHEATON®
15 - 415, gris
N° de catalogue 215-4042
Fournisseur: DWK Life Sciences
Aller à
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Spécifications
- Filetage:15 - 415
- Seq:35
- Couleur bouchon:gris