Bouchons compte-gouttes, WHEATON®
8 - 425, rouge
Fournisseur: DWK Life Sciences
Aller à
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Spécifications
- Filetage:8 - 425
- Seq:10
- Couleur bouchon:rouge