Trypsinizing flasks, WHEATON®
35 ml, Top screw cap 24-430, Screw cap 20-415
Codice catalogo 392-0369
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Borosilicate glass, clear.
- With pour out and caps
- Fluted design improves mixing
- Deep skirted screw caps
These flasks feature deep-fluted sides to promote mixing. A dam below the sidearm retains untrypsinized tissue when decanting. A deep skirted screw cap on the sidearm prevents contamination of flask contents. The sidearm allows easier decanting of contents while retaining large particles behind the dam. Sidearm caps are 20-415 size. Caps have white styrene-butadiene rubber liners.
Specifiche
- Capacité:35 ml
- Désignation:Top screw cap 24-430
- Col:Screw cap 20-415