Snap caps for sample bottles, WHEATON®
Snap caps for sample bottles, WHEATON®
30 mm, 16/24/30/37/44 ml
N° de catalogue WHEA242615
Fournisseur:   DWK Life Sciences
Snap caps for sample bottles, WHEATON®
N° de catalogue WHEA242615
Fournisseur:  DWK Life Sciences
Snap caps for sample bottles, WHEATON®
Snap caps for sample bottles, WHEATON®
30 mm, 16/24/30/37/44 ml
Prix
82.70 SFr.
Connexion Pour la disponibilité
1 * 200 ST
Product Details & Documents
Natural low density polyethylene.
  • All sizes have tabs for easy removal
  • Good chemical resistance
  • Good temperature resistance
For WHEATON snap cap sample bottles.
Spécifications
  • Taille du bouchon:
    30 mm
  • Pour flacons:
    16/24/30/37/44 ml