Bains cryothermostats circulation, srie Optima R, Grant, Ouverture l x p: 110x145 mm, Niveau de liquide min/max: 80/140 mm, GP2 00-R1, Gamme de temp.: -20...100 C, Prcision de temp.: 0.1 C 1 * 1 Pce
Catalog # 461-0213
Supplier: GRANT INSTRUMENTS
Frequently Bought Together