Filtres à membrane, type 184
184, 0,20 µm, 142 mm, -
SART18407-142-N
- Ø:142 mm
- Taille de pore:0,20 µm
- Stérile:-
- Type:184
- Seq:14
- Cat. No.:SART18407-142-N
- Épaisseur:170 µm
- Certification environnementale:Renewable Energy Certificate
- Max. temperature:180 °C
- Débit d'eau:15 ml/min/cm² (1 bar)
- Environmentally Preferable:Low Manufacturing Impact