Tween® 80 (Polysorbate) 10% en solution aqueuse
Flacon Verre, 25 mL
Katalognr. EURLEMR246025
Anbieter: EUROCLONE
Tween® 80 (Polysorbate) 10% en solution aqueuse
Katalognr. EURLEMR246025
Anbieter: EUROCLONE
CAS Nummer: 9005-65-6
Direkt zu
Product Details & Documents
- CAS Nummer:9005-65-6
- MDL-Nummer:MFCD00082107
- Lagertemperatur:Température ambiante
- EINECS:500-019-9
Technische Daten
Technische Daten
- Type de conditionnement:Flacon Verre
- Cdt:25 mL
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



