Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

TE buffer solution 100x concentrate (TRIS-EDTA buffer) pH 8,0, 1 KIT
Catalog # EURLEMR226001
Supplier:   EUROCLONE
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0
Catalog # EURLEMR226001
Supplier:  EUROCLONE
CAS Number:  
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

TE buffer solution 100x concentrate (TRIS-EDTA buffer) pH 8,0, 1 KIT

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications

Specifications

  • Cdt:
    1 KIT
  • Désignation:
    TE buffer solution 100x concentrate (TRIS-EDTA buffer) pH 8,0

Specifications