Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Streptavidin

Streptavidin

Flacon verre pour solides, 10 mg
Catalog # EURLEMR217010
Supplier:   EUROCLONE
Streptavidin
Catalog # EURLEMR217010
Supplier:  EUROCLONE
CAS Number:  
Streptavidin

Flacon verre pour solides, 10 mg

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 10 mg
Product Details & Documents
Specifications

Specifications

  • Type de conditionnement:
    Flacon verre pour solides
  • Cdt:
    10 mg
  • Cat. No.:
    EURLEMR217010
  • N ° CAS.:
    9013-20-1
  • Conditions de stockage:
    Freezer
  • MDL No.:
    MFCD00082035
  • Nom de la protéine/du peptide:
    Streptavidin

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.