Sodium hydroxyde
Sodium hydroxyde
5 kg
Catalog # EURLEMR212005
Corrosive
Supplier:   EUROCLONE
Sodium hydroxyde
Catalog # EURLEMR212005
Supplier:  EUROCLONE
CAS Number:  1310-73-2
Sodium hydroxyde
5 kg
CAS Number:  1310-73-2
Price
Inquire For Price
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2,13 g/cm³ (20 °C)
  • Storage Temperature:Température ambiante
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Specifications
Specifications
  • Cdt:
    5 kg
Specifications