Brij® 58
1 KIT
N° de catalogue EURLEMR103500
Fournisseur: EUROCLONE
Aller à
Product Details & Documents
Synonymes: Polyoxyethylene (20) cetyl ether, Polyethylene glycol hexadecyl ether
- Numéro CAS:9004-95-9
- Formule:C₅₆H₁₁₄O₂₁
- Numéro MDL:MFCD00080892
- Poids moléculaire:1123.51 g/mol
- EINECS:500-014-1
Spécifications
Spécifications
- Cdt:1 KIT
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



