Acide borique
5 kg
Catalog # EURLEMR100000
Supplier: EUROCLONE
Jump to:
Product Details & Documents
Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
- CAS Number:10043-35-3
- Formula:B(OH)₃
- MDL Number:MFCD00011337
- Molecular Weight:61.83 g/mol
- Merck Index:13,01326
- Boiling Pt:185 °C (1013 hPa)
- Melting Pt:160 °C
- Density:1,44 g/cm³ (25 °C)
- Storage Temperature:Température ambiante
- EINECS:233-139-2
- REACH:01-2119486683-25
Specifications
Specifications
- Cdt:5 kg



