Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Acide acétique

Acide acétique

Flacon Verre, 500 mL
Catalog # EURLEMR080500
Flammable
Corrosive
Supplier:   EUROCLONE
Acide acétique
Catalog # EURLEMR080500
Supplier:  EUROCLONE
CAS Number:  64-19-7
Acide acétique
Acide acétique

Flacon Verre, 500 mL

CAS Number:  64-19-7
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Vinegar acid, Methanecarboxylic acid, Ethanoic acid
  • CAS Number:64-19-7
  • Formula:H₃CCOOH
  • MDL Number:MFCD00036152
  • Molecular Weight:60.05 g/mol
  • Boiling Pt:118 °C (1013 hPa)
  • Melting Pt:17 °C
  • Density:1,05 g/cm³ (20 °C)
  • Flash Pt:38,5 °C
  • Storage Temperature:Température ambiante
  • EINECS:200-580-7
  • UN:2789
  • ADR:8,II
  • REACH:01-2119475328-30
Specifications

Specifications

  • Type de conditionnement:
    Flacon Verre
  • Cdt:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.