Hose clips, worm-threaded
12 - 20 mm
N° de catalogue BURK8677-1220
Fournisseur: BURKLE
Aller à
Product Details & Documents
Hose clip sleeve made of stainless steel 1,4016, screw zinc coated, with screw thread.
Documents
BURK8677-0812_TDS_EN BURK8677-1016_TDS_EN BURK8677-1220_TDS_EN
Fiche technique
Spécifications
- Largeur de serrage:12 - 20 mm
- Seq:03