Micro scissors for endoscopic work, Aesculap® MINOP®
Replacement handle for MINOP instruments FF385R - FF389R
N° de catalogue AESCFF432R
Fournisseur: Aesculap
Micro scissors for endoscopic work, Aesculap® MINOP®
N° de catalogue AESCFF432R
Fournisseur: Aesculap
Aller à
Product Details & Documents
Rigid micro scissors for advanced intraventricular Neuroendoscopy.
- Very delicate
Ø = 2 mm
Informations de commande: Complete instrument consists of a handle, an outer tube, and a jaw part with inner tube. Replacements can be ordered separately.
Spécifications
- Longueur:Replacement handle for MINOP instruments FF385R - FF389R