Micro scissors for endoscopic work, Aesculap® MINOP®
FF385R, sharp/sharp, 265 mm
N° de catalogue AESCFF385R
Fournisseur: Aesculap
Micro scissors for endoscopic work, Aesculap® MINOP®
N° de catalogue AESCFF385R
Fournisseur: Aesculap
Aller à
Product Details & Documents
Rigid micro scissors for advanced intraventricular Neuroendoscopy.
- Very delicate
Ø = 2 mm
Informations de commande: Complete instrument consists of a handle, an outer tube, and a jaw part with inner tube. Replacements can be ordered separately.
Spécifications
- Type:FF385R, sharp/sharp
- Longueur:265 mm
- Seq:02